missing translation for 'onlineSavingsMsg'
Learn More

KLRA1 Antibody, HRP, Novus Biologicals™

Product Code. 18416580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416580 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416580 Supplier Novus Biologicals Supplier No. NBP177343H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLRA1 Polyclonal antibody specifically detects KLRA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLRA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias killer cell lectin-like receptor subfamily A pseudogene 1, KLRA#, KLRA1, Ly49, Ly-49L, LY49L, MGC126520, MGC126522
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the center of human KLRA1. The immunogen is located within amino acids 140 - 190 of KLRA1. Amino Acid Squence: CYYFTMQKKDWKG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10748
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.