missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Klotho Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA129899
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLOTHO is the systemic anti-aging hormone within the glycosidase1 superfamily. It encodes a type I membrane protein that is abundant in the kidney and brain. In mice, a deficiency in KLOTHO expression leads to various systemic phenotypes resembling human aging such as arteriosclerosis, osteoporosis, and skin atrophy together with growth retardation, short life-span and infertility. Transgenic mice overexpressing KLOTHO have an extended life span by inhibiting insulin/IGF1 signaling. KLOTHO is involved in the regulation of calcium/phosphorus homeostasis by inhibiting the synthesis of active vitamin D and identified as a potential tumor suppressor.
Specifications
| Klotho | |
| Polyclonal | |
| Unconjugated | |
| KL | |
| alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16591 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| 0.1M tris HCl with 0.2% BSA and 0.05% sodium azide; pH 7.5 | |
| O35082 | |
| KL | |
| Synthetic peptide of 17 amino acids (Mouse) (C terminal). | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction