missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klotho beta Antibody (1025C) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB58891JF669
This item is not returnable.
View return policy
Description
Klotho beta Monoclonal antibody specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| Klotho beta | |
| Monoclonal | |
| Janelia Fluor 669 | |
| beta-klotho, betaKlotho, BKL, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| Mouse myeloma cell line, NS0-derived recombinant human Klotho β with a C-terminal 10 His tag., Phe53-Leu997, Accession # Q86Z14 | |
| RUO | |
| 152831 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 1025C | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction