missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Klotho Polyclonal specifically detects Klotho in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
Specifications
| Applications | ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Purification Method | Affinity Purified |
| Test Specificity | Three isoforms of KLOTHO are known to exist. |
| Target Species | Human, Mouse |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?