missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLLN Monoclonal antibody specifically detects KLLN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | KLLN |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | S08-5C6 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | CWS4, KILLIN, Killin, P53-Regulated DNA Replication Inhibitor |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human KLLN (Uniprot#: B2CW77) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?