missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLLN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546688
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: DSSGGKSSSS FARGALAWCR QRNPNPSCAA AETGARTSLP KERCRGWRLG Sequence homology: Human: 100%
KLLN Polyclonal antibody specifically detects KLLN in Human samples. It is validated for Western Blot.Specifications
KLLN | |
Polyclonal | |
Unconjugated | |
KLLN | |
CWS4; KILLIN; killin, p53-regulated DNA replication inhibitor; KLLN | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100144748 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B2CW77 | |
KLLN | |
Synthetic peptide directed towards the middle region of human KLLN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |