missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLLN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567349
This item is not returnable.
View return policy
Description
Immunogen sequence: SSSFARGALAW CRQRNPNPSC AAAETGARTS LPKERCRGWR LGNWLHKHPH PNTCPRLPAC WLPPILTERG ERVPKLVPLL Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 29%.
The protein encoded by this intronless gene is found in the nucleus, where it can inhibit DNA synthesis and promote S phase arrest coupled to apoptosis. The expression of this DNA binding protein is upregulated by transcription factor p53.
Specifications
| KLLN | |
| Polyclonal | |
| Unconjugated | |
| KLLN | |
| CWS4; KILLIN; killin, p53-regulated DNA replication inhibitor; KLLN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100144748 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B2CW77 | |
| KLLN | |
| Recombinant Human KLLN. Recombinant protein control fragment (Product #RP-108003). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction