missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK14, Mouse anti-Human, Clone: 2A7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00043847-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: SSPIARYPASLQCVNINISPDEVCQKAYPRTITPGMVCAGVPQGGKDSCQGDSGGPLVCRGQLQGLVSWGMERCALPGYPGVYTNLCKYRSWIEETMRDKSpecifications
| KLK14 | |
| Monoclonal | |
| Unconjugated | |
| kallikrein-related peptidase 14 | |
| KLK-L6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2A7 | |
| In 1x PBS, pH 7.4 | |
| NM_022046 | |
| KLK14 | |
| KLK14 (NP_071329.1, 152 a.a. ∼ 251 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 43847 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction