missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55394-1-AP-150UL
This item is not returnable.
View return policy
Description
KLHL9 is substrate-specific adapter for a CUL3-based E3 ubiquitin-protein ligase complex. Within this complex, it controls the dynamic behavior of AURKB on mitotic chromosomes and thereby coordinates faithful mitotic progression and completion of cytokinesis.Specifications
| KLHL9 | |
| Polyclonal | |
| Unconjugated | |
| KLHL9 | |
| kelch like 9 (Drosophila), Kelch like protein 9, KIAA1354, KLHL9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55958 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9P2J3 | |
| KLHL9 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction