missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553096
This item is not returnable.
View return policy
Description
Immunogen sequence: VISVEGKVYA VGGHDGNEHL GSMEMFDPLT NKWMMKASMN TKRRGIALAS LGGPIYAIGG LDDNTCFNDV ERYDIESDQW STVAPMNTPR GGVGSVALVN HVYAVGGNDG MASLSSVE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Kelch-like protein 8 is a protein that in humans is encoded by the KLHL8 gene. The mRNA contains a repetitive element in its 3-prime UTR, and the deduced 451-amino acid protein shares 36% identity with the actin-binding protein MAYVEN. RT-PCR ELISA detected highest expression in adult brain, liver, skeletal muscle, kidney, and ovary, and in several specific brain regions, predomitly amygdala and thalamus. Moderate expression was detected in all other adult and fetal tissues examined.
Specifications
| KLHL8 | |
| Polyclonal | |
| Unconjugated | |
| KLHL8 | |
| KIAA1378 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57563 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2G9 | |
| KLHL8 | |
| Recombit protein corresponding to Human KLHL8 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction