missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ KLHL7 293T Cell Transient Overexpression Lysate (Denatured) (T03)

Product Code. 16108576
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16108576 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16108576 Supplier Abnova™ Supplier No. H00055975T03.100uL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human overexpression lysate

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description KLHL7 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_001031710
Molecular Weight (g/mol) Theoretical M.W. 66kDa
Plasmid pCMV-KLHL7 full-length
Quality Control Testing Transient overexpression cell lysate was tested with anti-KLHL7 antibody (Mfr. No. H00055975-B01P) by Western Blot.
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 100UL KLHL7 293T Cell Transient Overexpression Lys
Signalwort
  • Warning
Gefahrenkategorie
  • Skin sensitiser Category 1
Gefahrenhinweis
  • H317-May cause an allergic skin reaction.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Mercaptoethanol
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.