missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL6 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-46128
This item is not returnable.
View return policy
Description
KLHL6 Polyclonal specifically detects KLHL6 in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| KLHL6 | |
| Polyclonal | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_569713.2 | |
| KLHL6 | |
| Peptide with sequence C-SERTKPRMHEFQSE corresponding to internal region according to NP_569713.2. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human and Mouse. Expected from sequence similarity: Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ00029, kelch-like 6 (Drosophila), kelch-like protein 6, kelch-like protein KLHL6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 89857 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction