missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL41 Monoclonal Antibody (1H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521008
This item is not returnable.
View return policy
Description
Peptide Sequence: RTDKENRVKN LSEVFDCIRF RLMTEKYFKD HVEKDDIIKS NPDLQKKIKV LKDAFAGKLP EPSKNAAKTG AGEVNGDVGD EDLLPGYLND IPRHGMFVKD.
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.
Specifications
| KLHL41 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O60662 | |
| KLHL41 | |
| KLHL41 (NP_006054.2, 205 a.a. ~ 304 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 1H3 | |
| Unconjugated | |
| KLHL41 | |
| Gm112; KBTBD10; kelch like family member 41; kelch related protein 1; kelch repeat and BTB (POZ) domain containing 10; kelch repeat and BTB domain-containing protein 10; kelch-like 41; kelch-like family member 41; kelch-like protein 41; Kelch-related protein 1; Kel-like protein 23; Klhl41; KRP1; sarcomeric muscle protein; Sarcosin | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10324 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction