missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL31 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24781-1-AP-150UL
This item is not returnable.
View return policy
Description
The function remains unknown.Specifications
| KLHL31 | |
| Polyclonal | |
| Unconjugated | |
| KLHL31 | |
| BKLHD6, KBTBD1, kelch like 31 (Drosophila), Kelch like protein 31, Kelch like protein KLHL, KLHL, KLHL31 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 244923, 401265 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BWA5, Q9H511 | |
| KLHL31 | |
| KLHL31 Fusion Protein Ag20462 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction