missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL29 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561867
This item is not returnable.
View return policy
Description
Immunogen sequence: SNCLGVLAMA EAMQCSELYH MAKAFALQIF PEVAAQEEIL SISKDDFIAY VSNDSLNTKA EELVYETVIK WIKKDPATRT QYAAELLAVV RLPFIHPSYL LNVV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
KLHL29 (Kelch-like protein 29), also known as KBTBD9, is a 655 amino acid protein that is related to the Drosophila Kelch protein. Mutations affecting Kelch function result in failure of Kelch to associate with the ring canals and subsequent female sterility. Human KLHL29 contains six kelch repeats and one BTB (POZ) domain. The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KLHL29 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 2p24.1.
Specifications
| KLHL29 | |
| Polyclonal | |
| Unconjugated | |
| KLHL29 | |
| A230106N14Rik; Gm68; KBTBD9; kelch like family member 29; kelch repeat and BTB (POZ) domain containing 9; kelch repeat and BTB domain-containing protein 9; kelch-like 29; kelch-like family member 29; kelch-like protein 29; Kiaa1921; Klhl29; mKIAA1921 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96CT2 | |
| KLHL29 | |
| Recombinant protein corresponding to Human KLHL29. Recombinant protein control fragment (Product #RP-96052). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction