missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL25 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67774-1-IG-20UL
This item is not returnable.
View return policy
Description
The KLHL25 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KLHL25. This antibody recognizes human, mouse antigen. The KLHL25 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| KLHL25 | |
| Monoclonal | |
| 1000 μg/mL | |
| ENC-2; ENC2; FLJ12587; FLJ45015 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 66 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D8B8 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 64410 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction