missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17082
This item is not returnable.
View return policy
Description
KLHL25 Polyclonal specifically detects KLHL25 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| KLHL25 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| ectodermal-neural cortex 2, ENC-2BTB/POZ KELCH domain protein, ENC2ectoderm-neural cortex protein 2, FLJ12587, FLJ45015, kelch-like 25 (Drosophila), Kelch-like protein 25 | |
| Rabbit | |
| 66 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Xenopus laevis (92%), Xenopus tropicalis (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.2mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9H0H3 | |
| KLHL25 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human KLHL25. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 64410 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction