missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16952-1-AP-20UL
This item is not returnable.
View return policy
Description
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for efficient chromosome alignment and cytokinesis. The BCR(KLHL21) E3 ubiquitin ligase complex regulates localization of the chromosomal passenger complex (CPC) from chromosomes to the spindle midzone in anaphase and mediates the ubiquitination of AURKB. Ubiquitination of AURKB by BCR(KLHL21) E3 ubiquitin ligase complex may not lead to its degradation by the proteasome.Specifications
| KLHL21 | |
| Polyclonal | |
| Unconjugated | |
| KLHL21 | |
| kelch like 21 (Drosophila), Kelch like protein 21, KIAA0469, KLHL21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 242785, 313743, 9903 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D4A2K4, Q3U410, Q9UJP4 | |
| Klhl21 | |
| KLHL21 Fusion Protein Ag10582 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction