missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLHL2 Monoclonal antibody specifically detects KLHL2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | KLHL2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI1G7 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ABP-KELCH, Actin-binding protein Mayven, kelch, kelch (Drosophila)-like 2 (Mayven), kelch-like 2, Mayven (Drosophila), kelch-like protein 2, MAV, MAYVEN |
| Host Species | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?