missing translation for 'onlineSavingsMsg'
Learn More

KLHL2 Antibody (OTI1G7), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18417677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417677 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417677 Supplier Novus Biologicals Supplier No. NBP271695AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLHL2 Monoclonal antibody specifically detects KLHL2 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone OTI1G7
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias ABP-KELCH, Actin-binding protein Mayven, kelch, kelch (Drosophila)-like 2 (Mayven), kelch-like 2, Mayven (Drosophila), kelch-like protein 2, MAV, MAYVEN
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11275
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.