missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107155
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 87%; Dog: 93%; Horse: 82%; Human: 100%; Pig: 81%; Rabbit: 93%; Rat: 81%; Yeast: 91% Immunogen sequence: GVGGQDGLGA PKDQQQPPQQ QPSQQQQPPP QEEPGTPSSS PDDKLLTSPR.
KLHL14 is a member of the KLHL family.
Specifications
| KLHL14 | |
| Polyclonal | |
| Unconjugated | |
| KLHL14 | |
| 6330403N15Rik; AI785250; kelch like family member 14; kelch-like 14; kelch-like family member 14; kelch-like protein 14; Kiaa1384; KLHL14; Printor; protein interactor of Torsin-1A; protein interactor of torsinA; RGD1566178; si:dkeyp-7a3.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57565 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9P2G3 | |
| KLHL14 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human KLHL14. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction