missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL13 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£375.00
Specifications
| Antigen | KLHL13 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18305966
|
Novus Biologicals
NBP3-04601-20UL |
20 μL |
N/A
|
N/A | |||||
|
18319336
|
Novus Biologicals
NBP3-04601-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLHL13 Polyclonal specifically detects KLHL13 in Human, Mouse samples. It is validated for Western Blot.Specifications
| KLHL13 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 90293 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| BTB and kelch domain containing 2, BTB and kelch domain-containing protein 2, kelch-like 13 (Drosophila), kelch-like protein 13, KIAA1309, MGC74791 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human KLHL13 (NP_277030.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title