missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL13 Monoclonal Antibody (8D1)

Description
MA5-15658 targets KLHL13 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15658 immunogen is purified recombinant fragment of human KLHL13 expressed in E. Coli. . MA5-15658 detects KLHL13 which has a predicted molecular weight of approximately 74kDa.
This gene encodes a BTB and kelch domain containing protein and belongs to the kelch repeat domain containing superfamily of proteins. The encoded protein functions as an adaptor protein that complexes with Cullin 3 and other proteins to form the Cullin 3-based E3 ubiquitin-protein ligase complex. This complex is necessary for proper chromosome segregation and completion of cytokinesis. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | KLHL13 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8D1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | KLHL13 |
| Gene Accession No. | Q80TF4, Q9P2N7 |
| Gene Alias | 1200009K10Rik; AI451128; Bklhd2; BTB and kelch domain containing 2; BTB and kelch domain-containing protein 2; D130072F20Rik; FLJ10262; kelch like family member 13; kelch-like 13; kelch-like family member 13; kelch-like protein 13; Kiaa1309; KLHL13; MGC74791; mKIAA1309 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?