Learn More
Invitrogen™ KLHL12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143992
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human HEK293 whole cell, human Hela whole cell, human Jurkat whole cell. IHC: human hashimoto thyroiditis tissue, human liver cancer tissue, human lung cancer tissue, human thyroid papillary carcinoma tissue, mouse brain tissue, rat brain tissue, rat lymph node tissue. ICC/IF: T47D cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.
Specifications
| KLHL12 | |
| Polyclonal | |
| Unconjugated | |
| Klhl12 | |
| C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 240756, 266772, 59349 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q53G59, Q8BZM0, Q8R2H4 | |
| Klhl12 | |
| E.coli-derived human KLHL12 recombinant protein (Position: R27-I331). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.