missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL12/C3IP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | KLHL12/C3IP1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KLHL12/C3IP1 Polyclonal specifically detects KLHL12/C3IP1 in Rat samples. It is validated for Western Blot.Specifications
| KLHL12/C3IP1 | |
| Polyclonal | |
| Rabbit | |
| NP_714952 | |
| 59349 | |
| Synthetic peptide towards C3IP1. Peptide sequence DIMTNTHAKSILNSMNSLRKSNTLCDVTLRVEQKDFPAHRIVLAACSDYF. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1 | |
| KLHL12 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title