missing translation for 'onlineSavingsMsg'
Learn More

KLHL12/C3IP1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18406324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406324 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406324 Supplier Novus Biologicals Supplier No. NBP149927AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLHL12/C3IP1 Polyclonal antibody specifically detects KLHL12/C3IP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL12/C3IP1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Kelch-Like 12 using the numbering given in entry NP_067646.1 (GeneID 59349).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 59349
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.