Learn More
Invitrogen™ KLHL11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139816
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YSIGGHGNFS PGFKDVTVYN PELDKWHNLE SAPKILRDVK ALAIEDRFVY IAARTPVDRD TEDGLKAVIT CYDTETRQWQ DVESLPLIDN YCFFQMSVVN S Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 99%.
Kelch-like protein 11, Subcellular location Secreted.
Specifications
| KLHL11 | |
| Polyclonal | |
| Unconjugated | |
| KLHL11 | |
| BC011167; FLJ10572; kelch like family member 11; kelch-like 11; kelch-like family member 11; kelch-like protein 11; KLHL11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55175 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVR0 | |
| KLHL11 | |
| Recombinant protein corresponding to Human KLHL11. Recombinant protein control fragment (Product #RP-108892). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.