missing translation for 'onlineSavingsMsg'
Learn More

KLHL1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18404047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404047 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404047 Supplier Novus Biologicals Supplier No. NBP176670DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLHL1 Polyclonal antibody specifically detects KLHL1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ30047, kelch (Drosophila)-like 1, kelch-like 1 (Drosophila), KIAA1490kelch-like protein 1, Mayven-related protein 2, MRP2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the amino-terminus of human KLHL1. The immunogen is located within the first 50 amino acids of KLHL1. Amino Acid Squence: MSGSGRKDFDVKHI
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57626
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.