Learn More
Invitrogen™ KLHDC8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113869
Description
Immunogen sequence: AMAEGSVFSL GGLQQPGPHN FYSRPHFVNT VEMFDLEHGS WTKLPRSLRM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Specifications
| KLHDC8B | |
| Polyclonal | |
| Unconjugated | |
| KLHDC8B | |
| 4931406O17Rik; CHL; FLJ11302; FP17659; kelch domain containing 8B; kelch domain-containing protein 8B; KLHDC8B; MGC35097 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 200942 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IXV7 | |
| KLHDC8B | |
| Synthetic peptide directed towards the middle region of human KLHDC8B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.