missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHDC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04568-20UL
This item is not returnable.
View return policy
Description
KLHDC2 Polyclonal specifically detects KLHDC2 in Human samples. It is validated for Western Blot.
Specifications
| KLHDC2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23588 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| HCA33, HCLP-1Host cell factor-like protein 1, Hepatocellular carcinoma-associated antigen 33, Host cell factor homolog LCP, kelch domain containing 2, kelch domain-containing protein 2, LCP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human KLHDC2 (NP_055130.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction