missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLH18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557121
This item is not returnable.
View return policy
Description
Immunogen sequence: EAKDYHLMPE RRPHLPAFRT RPRCCTSIAG LIYAVGGLNS AGDSLNVVEV FDPIANCWER CRPMTTARSR VGVAVVNGL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
KLHL18 is a protein coding gene.
Specifications
| KLH18 | |
| Polyclonal | |
| Unconjugated | |
| KLHL18 | |
| A930041K15; AW545966; kelch like family member 18; kelch-like 18; kelch-like family member 18; kelch-like protein 18; KIAA0795; Klhl18; OK/SW-cl.74; RGD1310570 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94889 | |
| KLHL18 | |
| Recombinant protein corresponding to Human KLH18. Recombinant protein control fragment (Product #RP-96311). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction