missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLH Antibody, Texas Red, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97387
This item is not returnable.
View return policy
Description
KLH Polyclonal specifically detects KLH in All Species samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| KLH | |
| Polyclonal | |
| Texas Red | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum. | |
| All species | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2500 | |
| keyhole limpet hemocyanin (KLH) | |
| KLH (Keyhole Limpet Hemocyanin) | |
| 0.1 mL | |
| Primary | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction