missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLH Polyclonal specifically detects KLH in All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | KLH |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:10-1:500, Fluorophore-linked immunosorbent assay |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Alias | keyhole limpet hemocyanin (KLH) |
| Host Species | Rabbit |
| Immunogen | KLH (Keyhole Limpet Hemocyanin) |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?