missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLH Polyclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | Janelia Fluor 646 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | keyhole limpet hemocyanin (KLH) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant KLH (Keyhole Limpet Hemocyanin). |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?