missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLH Polyclonal specifically detects KLH in Non-species specific samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | KLH |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | keyhole limpet hemocyanin (KLH) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Keyhole Limpet Hemocyanin. |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?