missing translation for 'onlineSavingsMsg'
Learn More

KLH Antibody (15F7E4G3), Janelia Fluor™ 549 , Novus Biologicals™

Product Code. 18418208 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418208 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418208 Supplier Novus Biologicals Supplier No. NBP305779JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLH Monoclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLH
Applications Western Blot, ELISA
Classification Monoclonal
Clone 15F7E4G3
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias keyhole limpet hemocyanin (KLH)
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant KLH (Keyhole Limpet Hemocyanin).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.