missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5117090
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Positive Samples: A-549, K-562, HepG2, Mouse brain, Mouse kidney, Rat brain Immunogen sequence: MSAAAYMDFV AAQCLVSISN RAAVPEHGVA PDAERLRLPE REVTKEHGDP GDTWKDYCTL VTIAKSLLDL NKYRPIQTPS VCSDSLESPD EDMGSDSDVT TESGSSPSHS PEERQDPGSA PSPLSLLHPG VAAKGKHASE KRHKCPYSGC.
KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregnancy and tumorigenesis.
Specifica
| KLF9 | |
| Polyclonal | |
| Unconjugated | |
| KLF9 | |
| 2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117560, 16601, 687 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.86 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O35739, Q01713, Q13886 | |
| KLF9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human KLF9 (NP_0011971). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto