missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107154
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregnancy and tumorigenesis.
Specifications
| KLF9 | |
| Polyclonal | |
| Unconjugated | |
| KLF9 | |
| 2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117560, 687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q01713, Q13886 | |
| KLF9 | |
| Synthetic peptide directed towards the N-terminal region of Human KLF9. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction