missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577883
This item is not returnable.
View return policy
Description
Positive Control: A431, HeLa Predicted Reactivity: Mouse (97%), Rat (98%), Pig (98%), Rhesus Monkey (97%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregnancy and tumorigenesis.
Specifications
| KLF9 | |
| Polyclonal | |
| Unconjugated | |
| KLF9 | |
| 2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q13886 | |
| KLF9 | |
| Recombinant protein encompassing a sequence within the center region of human KLF9. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction