missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27418SS
This item is not returnable.
View return policy
Description
KLF8 Polyclonal specifically detects KLF8 in Human, Primate samples. It is validated for Western Blot.
Specifications
| KLF8 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Basic krueppel-like factor 3, basic kruppel-like factor 3, BKLF3DKFZp686O08126, DXS741, Krueppel-like factor 8, Kruppel-like factor 8, Zinc finger protein 741, ZNF741MGC138314 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11279 | |
| Human, Primate | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml | |
| O95600 | |
| KLF8 | |
| A portion of amino acids 1-50 from human kLF8 was used as the immunogen for this antibody. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, sumatran orangutan, and white-cheeked gibbon, 89% in rhesus monkey, and 78% in new world monkey. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction