missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27418
This item is not returnable.
View return policy
Description
KLF8 Polyclonal specifically detects KLF8 in Human, Primate samples. It is validated for Western Blot.
Specifications
| KLF8 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Basic krueppel-like factor 3, basic kruppel-like factor 3, BKLF3DKFZp686O08126, DXS741, Krueppel-like factor 8, Kruppel-like factor 8, Zinc finger protein 741, ZNF741MGC138314 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11279 | |
| Human, Primate | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml | |
| O95600 | |
| KLF8 | |
| A portion of amino acids 1-50 from human kLF8 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, sumatran orangutan, and white-cheeked gibbon, 89% in rhesus monkey, and 78% in new world monkey. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido