missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101340
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KLF7.
The protein encoded by this gene is a member of the Kruppel-like transcriptional regulator family. Members in this family regulate cell proliferation, differentiation and survival and contain three C2H2 zinc fingers at the C-terminus that mediate binding to GC-rich sites. This protein may contribute to the progression of type 2 diabetes by inhibiting insulin expression and secretion in pancreatic beta-cells and by deregulating adipocytokine secretion in adipocytes. A pseudogene of this gene is located on the long arm of chromosome 3. Alternative splicing results in multiple transcript variants.
Specifications
| KLF7 | |
| Polyclonal | |
| Unconjugated | |
| Klf7 | |
| 9830124P08Rik; Klf7; Krueppel-like factor 7; Kruppel like factor 7; Kruppel-like factor 7 (ubiquitous); Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor; UKLF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8609, 93691 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75840, Q99JB0 | |
| Klf7 | |
| A synthesized peptide derived from human KLF7(Accession O75840), corresponding to amino acid residues V178-K228. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction