missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF7 Antibody (3G3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42626AF700
This item is not returnable.
View return policy
Description
KLF7 Monoclonal specifically detects KLF7 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| KLF7 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KLF7 | |
| Recombinant human KLF7 (1-302aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3G3 | |
| Western Blot, ELISA | |
| Krueppel-like factor 7, Kruppel-like factor 7 (ubiquitous), ubiquitous Kruppel-like transcription factor, UKLFUbiquitous krueppel-like factor | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8609 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction