missing translation for 'onlineSavingsMsg'
Learn More

KLF6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£176.00 - £393.00

Specifications

Antigen KLF6
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18021846
View Documents
Novus Biologicals
NBP3-05013-20UL
20 μL
£176.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18301137
View Documents
Novus Biologicals
NBP3-05013-100UL
100 μL
£393.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KLF6 Polyclonal specifically detects KLF6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

KLF6
Polyclonal
Purified
RUO
Human, Mouse
BCD1Zf9, B-cell-derived protein 1, COPEBCBA1, core promoter element binding protein, Core promoter element-binding protein, CPBPZF9, DKFZp686N0199, EC 2.1.1.43, EC 6.1.1.15, GBF, GC-rich binding factor, GC-rich sites-binding factor GBF, Krueppel-like factor 6, Kruppel-like factor 6, Kruppel-like zinc finger protein Zf9, PAC1, Proto-oncogene BCD1, protooncogene B-cell derived 1, ST12, suppression of tumorigenicity 12 (prostate), Suppressor of tumorigenicity 12 protein, Transcription factor Zf9
Recombinant fusion protein containing a sequence corresponding to amino acids 40-200 of human KLF6 (NP_001291.3).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Prostate Cancer
PBS with 50% glycerol, pH7.3.
1316
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.