missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF5 Recombinant Rabbit Monoclonal Antibody (HL2269)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550468
This item is not returnable.
View return policy
Description
KLF5 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.
Specifications
| KLF5 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q13887, Q9Z0Z7 | |
| KLF5 | |
| Recombinant fragment of human KLF5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2269 | |
| Unconjugated | |
| KLF5 | |
| (intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; basic transcription element-binding protein 2; BTEB2; BTE-binding protein 2; CKLF; colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; IKLF; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; transcription factor BTEB2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12224, 688, 84410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction