missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF5 Recombinant Superclonal™ Antibody (23HCLC)
KLF5 Recombinant Polyclonal Antibody (23HCLC)
Brand: Invitrogen 710918
This item is not returnable.
View return policy
Description
KLF5 Recombinant Polyclonal Antibody for ICC/IF, ChIP
Specifications
| KLF5 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q13887 | |
| KLF5 | |
| Peptides corresponding to Human KLF5 (aa 11-24, 358-372). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, Immunocytochemistry | |
| 23HCLC | |
| Unconjugated | |
| KLF5 | |
| (intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; basic transcription element-binding protein 2; BTEB2; BTE-binding protein 2; CKLF; colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; IKLF; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; transcription factor BTEB2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 688 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction