missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLF5 Polyclonal specifically detects KLF5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | KLF5 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Basic transcription element-binding protein 2, BTEB2, BTE-binding protein 2, CKLFGC-box-binding protein 2, Colon krueppel-like factor, colon kruppel-like factor, GC box binding protein 2, IKLFbasic transcription element binding protein 2, Intestinal-enriched krueppel-like factor, intestinal-enriched kruppel-like factor, Krueppel-like factor 5, Kruppel-like factor 5 (intestinal), Transcription factor BTEB2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human KLF5. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?