missing translation for 'onlineSavingsMsg'
Learn More

KLF5 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18387513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387513 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387513 Supplier Novus Biologicals Supplier No. NBP298939V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLF5 Polyclonal specifically detects KLF5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF5
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Basic transcription element-binding protein 2, BTEB2, BTE-binding protein 2, CKLFGC-box-binding protein 2, Colon krueppel-like factor, colon kruppel-like factor, GC box binding protein 2, IKLFbasic transcription element binding protein 2, Intestinal-enriched krueppel-like factor, intestinal-enriched kruppel-like factor, Krueppel-like factor 5, Kruppel-like factor 5 (intestinal), Transcription factor BTEB2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human KLF5.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 688
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.