missing translation for 'onlineSavingsMsg'
Learn More

KLF4 Polyclonal Antibody, Invitrogen™

Product Code. 13491247
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13491247 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13491247 Supplier Invitrogen Supplier No. PA527440

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA5-27440 targets KLF4 in FACS and WB applications and shows reactivity with Human and mouse samples. The PA5-27440 immunogen is recombit protein fragment corresponding to a region within amino acids 349 and 457 of Human KLF4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Transcription factor which acts as both an activator and repressor. Binds the CACCC core sequence. Binds to multiple sites in the 5′-flanking region of its own gene and can activate its own transcription. Required for establishing the barrier function of the skin and for postnatal maturation and maintece of the ocular surface. Involved in the differentiation of epithelial cells and may also function in skeletal and kidney development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF4
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Gene KLF4
Gene Accession No. O43474, Q60793
Gene Alias EZF, GKLF, Krueppel-like factor 4, endothelial Kruppel-like zinc finger protein, epithelial zinc finger protein EZF, gut-enriched krueppel-like factor
Gene Symbols KLF4
Host Species Rabbit
Immunogen Recombit protein fragment corresponding to a region within amino acids 349 and 457 of Human KLF4
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16600, 9314
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.