Learn More
KLF4 Antibody (CL5785) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61427JF646
Product Code. 30304554
  • £558.00 / 0.10mL
Description
KLF4 Monoclonal antibody specifically detects KLF4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KLF4
Monoclonal
Janelia Fluor 646
Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
This antibody was developed using a synthetic peptide derived from O43474, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Cycle and Replication
9314
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL5785
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified